Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JAB1 Antibody (OTI2B12), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271659G
Description
JAB1 Monoclonal specifically detects JAB1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
JAB1 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
COPS5 | |
Full length recombinant protein of human JAB1 (NP_006828) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI2B12 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
COP9 constitutive photomorphogenic homolog subunit 5 (Arabidopsis), COP9 signalosome complex subunit 5, CSN5COP9 (constitutive photomorphogenic, Arabidopsis, homolog) subunit 5, EC 3.4, JAB138 kDa Mov34 homolog, Jun activation domain-binding protein 1, MOV-34, SGN5MGC3149, Signalosome subunit 5 | |
Mouse | |
Affinity Purified | |
Signal Transduction | |
10987.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction