Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ JAB1 Monoclonal Antibody (3F10B10)
GREENER_CHOICE

Catalog No. 371500
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-150-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 37-150-0 Supplier Invitrogen™ Supplier No. 371500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

JAB1 Monoclonal Antibody for Western Blot, IP, ELISA

JAB-1 is a 38 kDa protein which was originally described as a coactivator for cJUN. It is present in a large protein complex called the Signalosome, which has been proposed to play a signaling role in mammalian cells. This protein complex appears to be analogous to the COP9 complex in plants, which is important in light-mediated plant development. Although the specific function of JAB-1 in this complex is unclear, it has recently been implicated in targeting proteins for degradation by the proteasome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JAB1
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 3F10B10
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene Cops5
Gene Accession No. O35864, Q92905
Gene Alias 38 kDa Mov34 homolog; AI303502; COP9 (constitutive photomorphogenic) homolog, subunit 5; COP9 (constitutive photomorphogenic) homolog, subunit 5 (Arabidopsis thaliana); COP9 complex S5; COP9 constitutive photomorphogenic homolog subunit 5; COP9 signalosome complex subunit 5; COP9 signalosome subunit 5; Cops5; CSN5; JAB1; Jun activation domain-binding protein 1; Jun coactivator; Kic2; Kip1 C-terminus-interacting protein 2; MGC3149; Mov34; MOV-34; SGN5; signalosome subunit 5; testis secretory sperm-binding protein Li 231m
Gene Symbols Cops5
Host Species Mouse
Immunogen Synthetic peptide derived from the internal region of JAB1.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10987, 26754
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.