Learn More
JAC Medical Phlebotomy Lab Trays
Description
- Green high-impact polystyrene
- Large lab tray carries up to 80 tubes and many other components
- Compact lab tray has a medium capacity for up to 60 tubes and selected components
Order Info
Handles sold separately: 22-038-419 (JAC 8); 22-038-420 (JAC 9). Lid not included.
Specifications
Specifications
| Autoclavable | Autoclavable |
| Holds | Up to 80 tubes |
| Product Type | Phlebotomy Lab Tray |
| Size | Large |
| For Use With (Application) | For single-purpose |
| Color | White |
| Material | Polystyrene |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.