Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DenLine™ Uniforms Protection Plus™ II DL247 Unisex Short Length Lab Jackets
SDP

Catalog No. 22412365
Click to view available options
Color:
Black
Royal Blue
White
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Lab coat provides AAMI PB70 Level 1 fluid resistance and offers protection from aerosols, blood, and all aqueous liquids.

  • Primary Applications:
    • Anatomic Pathology
    • Clinical Pathology
    • Clinical Laboratory
    • Phlebotomy
    • Specimen Collection
    • Blood Collection, Testing
    • Manufacturing
    • Dental
    • Human and Animal Research
    • Cleanroom ISO 5/Class 100 - Medical Device, Pharma, Electronics
  • AAMI PB70 Level 1 Protection
  • Provides wearers 5 ft. 2 in. and shorter frontal coverage to knees for OSHA 29 CFR 1910 compliance
  • Provides all height wearers protection to ankles when worn in combination with DenLine style DL385 UltraLite Unisex Drawstring Pant
  • Overall Air Permeability 45 CFM (ASTM D737)
  • Round neck, no knit ribbing to soil, fray or wear out prematurely
  • Stain resistant fabric and cuffs (blood, bleach and ALL aqueous reagents and liquids)
  • Anti-static fabric prevents false instrument readings, damage to computers
  • Lint free fabric prevents sample contamination, ideal for molecular testing
  • Durable construction, 3 Pockets, Stainless snap closures
  • Cuffs for overgloving
  • Bleach safe all colors, choice of 4 colors white, ceil blue, royal, and black
  • Autoclavable Fabric, 225 commercial wash tested

Specifications

Product Type General Purpose Lab Coat
Size 3X-Large
Gender Unisex
Material Polyester, Carbon
Color Royal Blue
Closure Type Snaps
Cuff Style Knit
Disposable Reusable
Sterility Non-sterile
Resistance Fluids, Static, Stains
Style Short
No. of Pockets 3
Collar Type Round
Latex Free Yes
Size (English) Chest 54 to 56 in.
Size (English) Hip 50 to 53.5 in.
Size (Metric) Chest 137 to 142 cm
Size (Metric) Hip 127 to 136 cm
Certifications/Compliance OSHA 29 CFR 1910, AAMI PB70 Level 1 Protection
For Use With (Application) Anatomic Pathology, Clinical Pathology, Clinical Laboratory, Phlebotomy, Specimen Collection, Research, Dental, Cleanrooms
Length (English) 31.5 in.
Length (English) Cuff 4 in.
Length (English) Sleeve 44 in.
Length (Metric) 80 cm
Length (Metric) Cuff 10.1 cm
Weight (English) 0.6 lb.
Autoclavable Autoclavable
Permeability Overall Air Permeability 45 CFM (ASTM D737)
Construction 360° Fluid Resistance Front, Shoulders, and Back
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.