Learn More
Ansell KleenGuard™ Nemesis™ Safety Glasses
GREENER_CHOICE
Catalog No. 19066116 Shop All Ansell Products
Change view
Click to view available options
Frame Color:
Metallic Blue
Red
Silver
Lens Tint:
Clear
Smoke
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint
19-066-116 Silver Smoke
19-066-113 Metallic Blue Clear
19-066-114 Metallic Blue Smoke
19-066-115 Silver Clear
19-066-117 Red Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-066-116 Supplier Ansell Supplier No. 47383
Add to Cart
Add to Cart
This item is not returnable. View return policy
KleenGuard Nemesis Safety Eyewear's stylish good looks and durable, lightweight construction are just some of the reasons why millions of people wear them for protection both at work and at home.
  • Sporty, flexible, lightweight design for comfort and compliance
  • Patented FLEX-DRY™ nosepiece design with integrated ridges to divert sweat
  • Every pair includes neck cord for reduced risk of damage when not being worn
  • Smoke lenses are worn on bright, sunny days to provide glare reduction; ideal for outdoor use
  • Anti-Fog coating to prevent fogging; ideal for humid environments, temperature variances
  • Hardcoated lenses for scratch resistance and longer life
  • Lenses are made of polycarbonate material that provides 99.9% UVA/UVB/UVC protection
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog
Certifications/Compliance ANSI Z87.1+2015
Lens Tint Smoke
Product Type Protective Glasses
For Use With (Application) Used for outdoor viewing
Frame Color Silver
Includes Neck cord
No. per Case 12/Cs.
Lens Material Polycarbonate
Frame Material Metal
UV Protection Yes (UVA, UVB, UVC)
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.