Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Jagged 1 Antibody (188331) [DyLight 405], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB1277E
Description
Jagged 1 Monoclonal antibody specifically detects Jagged 1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, CyTOFSpecifications
Jagged 1 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Flow Cytometry, CyTOF | |
188331 | |
Flow Cytometry, CyTOF-ready | |
AGS, AHDMGC104644, Alagille syndrome, AWS, CD339, CD339 antigen, HJ1, jagged 1, Jagged1, JAGL1, protein jagged-1 | |
NS0-derived recombinant human Jagged 1, Ser32-Asp296, Accession # P78504 | |
0.1 mL | |
Angiogenesis, Cancer, Growth and Development, Neuronal Cell Markers, Stem Cell Signaling Pathway | |
182 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction