Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Jagged 2 Antibody (746513), PE/Cy5.5, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB4748PECY55
Description
Jagged 2 Monoclonal antibody specifically detects Jagged 2 in Mouse samples. It is validated for Flow CytometrySpecifications
Jagged 2 | |
Monoclonal | |
PE-Cyanine5.5 | |
PBS | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry | |
746513 | |
Flow Cytometry | |
HJ2, jagged 2, Jagged2, protein jagged-2, SER2 | |
Mouse myeloma cell line NS0-derived recombinant mouse Jagged 2, Met27-Leu1084, Accession # Q9QYE5 | |
0.1 mL | |
Cell Cycle and Replication, Stem Cell Signaling Pathway | |
3714 | |
Store at 4C in the dark. Do not freeze. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction