Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Jagged1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA582765
Description
Immunogen sequence: LRKRRKPGSH THSASEDNTT NNVREQLNQI KNPIEKHGAN TVPIKDYENK NSKMSKIRTH NSEVEEDDMD KHQQKARFAK QPAYTLVDRE EKPPNGTPTK HPNWTNKQDN RDLESAQSLN RMEYI.
The jagged 1 protein encoded by JAG1 is the human homolog of the Drosophilia jagged protein. Human jagged 1 is the ligand for the receptor notch 1, the latter a human homolog of the Drosophilia jagged receptor notch. Mutations that alter the jagged 1 protein cause Alagille syndrome. Jagged 1 signalling through notch 1 has also been shown to play a role in hematopoiesis.
Specifications
Jagged1 | |
Polyclonal | |
Unconjugated | |
JAG1 | |
ABE2; AGS; AHD; AWS; CD339; Gsfabe2; HJ1; Htu; JAG1; jagged 1; jagged canonical Notch ligand 1; Jagged1; JAGL1; Ozz; protein jagged-1; Ser-1; Serrate-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
182 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P78504 | |
JAG1 | |
Recombinant protein corresponding to Human JAG1. Recombinant protein control fragment (Product #RP-93722). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction