Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Jagged1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
$191.80 - $393.00
Specifications
Antigen | Jagged1 |
---|---|
Concentration | 0.5 mg/mL |
Applications | ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FEPA5145348
|
Invitrogen™
PA5145348 |
20 μL |
Each for $191.80
|
|
|||||
FEPA5145347
|
Invitrogen™
PA5145347 |
200 μL |
Each for $393.00
|
|
|||||
Description
For immunocytochemistry the cells were formalin fixed.
The jagged 1 protein encoded by JAG1 is the human homolog of the Drosophilia jagged protein. Human jagged 1 is the ligand for the receptor notch 1, the latter a human homolog of the Drosophilia jagged receptor notch. Mutations that alter the jagged 1 protein cause Alagille syndrome. Jagged 1 signalling through notch 1 has also been shown to play a role in hematopoiesis.Specifications
Jagged1 | |
ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P78504, Q63722, Q9QXX0 | |
16449, 182, 29146 | |
Recombinant Human JAG1 protein, Gly470-Thr834 (Accession #P78504). | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
ABE2; AGS; AHD; AWS; CD339; Gsfabe2; HJ1; Htu; JAG1; jagged 1; jagged canonical Notch ligand 1; Jagged1; JAGL1; Ozz; protein jagged-1; Ser-1; Serrate-1 | |
JAG1 | |
IgG | |
Protein A | |
JAG1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title