Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Jagged1 Rabbit anti-Human, Clone: JF96-06, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP266912
Description
Jagged 1 Monoclonal specifically detects Jagged 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Jagged1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:100-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
AGS, AHDMGC104644, Alagille syndrome, AWS, CD339, CD339 antigen, HJ1, jagged 1, Jagged1, JAGL1, protein jagged-1 | |
Rabbit | |
Protein A purified | |
Angiogenesis, Cancer, Growth and Development, Neuronal Cell Markers, Stem Cell Signaling Pathway | |
182 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
JF96-06 | |
Unconjugated | |
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
JAG1 | |
Synthetic peptide within Human Jagged 1 aa 1182-1214/ 1218. (SwissProt: P78504 Human; SwissProt: Q9QXX0 Mouse; SwissProt: Q63722 Rat) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction