Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Jagged2 Monoclonal Antibody (K.902.6)

Rabbit Monoclonal Antibody
Supplier: Invitrogen™ MA514979
Description
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with Jagged1.
The Notch signaling pathway is an intercellular signaling mechanism that is essential for proper embryonic development. Members of the Notch gene family encode transmembrane receptors that are critical for various cell fate decisions. JAG2 is one of several ligands that activate Notch and related receptors.
Specifications
Jagged2 | |
Monoclonal | |
50 μg/mL | |
0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5 | |
Q9Y219 | |
Jag2 | |
Synthetic peptide corresponding to residues surrounding Pro1212 of human Jagged2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunoprecipitation, Western Blot | |
K.902.6 | |
Unconjugated | |
Jag2 | |
D12Ggc2e; HJ2; JAG2; jagged 2; Jagged2; mJagged2-1; protein jagged-2; SER2; Serh; sm; syndactylism | |
Rabbit | |
Affinity chromatography | |
RUO | |
3714 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction