Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Jagged2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5111319
Description
Immunogen sequence: LTLHFNRDHV PQGTTVGAIC SGIRSLPATR AVARDRLLVL LCDRASSGAS AVEVAVSFSP ARDLPDSSLI QGAAHAIVAA ITQRGNSSLL LAVTEVKVET VVTG.
The Notch signaling pathway is an intercellular signaling mechanism that is essential for proper embryonic development. Members of the Notch gene family encode transmembrane receptors that are critical for various cell fate decisions. JAG2 is one of several ligands that activate Notch and related receptors.
Specifications
| Jagged2 | |
| Polyclonal | |
| Unconjugated | |
| Jag2 | |
| D12Ggc2e; HJ2; JAG2; jagged 2; Jagged2; mJagged2-1; protein jagged-2; SER2; Serh; sm; syndactylism | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3714 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.20 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y219 | |
| Jag2 | |
| Recombinant Protein corresonding to Human Jagged2. Recombinant protein control fragment (Product #RP-106291). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction