Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Jak1 Antibody (413104), DyLight 550, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB4260L
Description
Jak1 Monoclonal antibody specifically detects Jak1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOFSpecifications
Jak1 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Simple Western, Immunohistochemistry, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF | |
413104 | |
Western Blot, Simple Western, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
EC 2.7.10, JAK-1, JAK1Atyrosine-protein kinase JAK1, JAK1B, Janus kinase 1EC 2.7.10.2, JTK3 | |
E. coli-derived recombinant human Jak1, Pro32-Phe286, Accession # P23458 | |
0.1 mL | |
Protein Kinase, Signal Transduction | |
3716 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction