Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Jak1 Antibody (7A5), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Jak1 |
---|---|
Clone | 7A5 |
Dilution | ELISA, Immunohistochemistry 1:50-1:200 |
Applications | ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
Jak1 Monoclonal antibody specifically detects Jak1 in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
Jak1 | |
ELISA, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
EC 2.7.10, JAK-1, JAK1Atyrosine-protein kinase JAK1, JAK1B, Janus kinase 1EC 2.7.10.2, JTK3 | |
A synthesized peptide derived from Human Jak1 [UniProt P23458] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
7A5 | |
ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Phospho Specific, Protein Kinase, Signal Transduction | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
3716 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title