Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Jak1, p Tyr1034 Antibody (SR1138), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32161050UL
Description
Jak1 Monoclonal antibody specifically detects Jak1 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
Jak1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
SR1138 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:100 | |
EC 2.7.10, JAK-1, JAK1Atyrosine-protein kinase JAK1, JAK1B, Janus kinase 1EC 2.7.10.2, JTK3 | |
A synthesized peptide derived from human phospho-Jak1 [Tyr1034, Tyr1035] (Uniprot #: P23458) | |
50 μg | |
Phospho Specific, Protein Kinase, Signal Transduction | |
3716 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction