Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ JAK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109871
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109871 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109871 Supplier Invitrogen™ Supplier No. PA5109871
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VSEKKPATEV DPTHFEKRFL.

Janus kinase 1 (JAK1), is a protein tyrosine kinase that is involved in the interferon-alpha/beta and -gamma signal transduction pathways. Immunological stimuli, such as interferons and cytokines, induce recruitment of Stat transcription factors to cytokine receptor-associated JAK1. JAK1 then phosphorylates proximal Stat factors, which subsequently dimerize, translocate to the nucleus and bind to cis elements upstream of target gene promoters to regulate transcription. Upon ligand binding, JAK1 undergoes tyrosine phosphorylation and catalytic activation in an interdependent manner. Phosphorylation of tyrosine residues at positions 1022 and 1023 is believed to function in the activation of catalytic events.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JAK1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.42 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene JAK1
Gene Accession No. P23458, P52332
Gene Alias AA960307; BAP004; C130039L05Rik; EC 2.7.10.2; Jak1; JAK-1; JAK1A; JAK1B; Janus kinase 1; Janus protein tyrosine kinase 1; JTK3; kinase Jak1; Tyrosine-protein kinase JAK1
Gene Symbols JAK1
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16451, 3716, 84598
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.