Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Jak1 Rabbit anti-Human, Mouse, Rat, Clone: JM75-03, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP275547
Description
Jak1 Monoclonal specifically detects Jak1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Jak1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500-1:1000, Flow Cytometry 1:50-1:100, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50-1:500 | |
EC 2.7.10, JAK-1, JAK1Atyrosine-protein kinase JAK1, JAK1B, Janus kinase 1EC 2.7.10.2, JTK3 | |
Rabbit | |
133 kDa | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
JM75-03 | |
Unconjugated | |
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
JAK1 | |
Synthetic peptide within Human Jak1 aa 621-670/ 1,154. (SwissProt: P23458 Human; SwissProt: P52332 Mouse; SwissProt: G3V9W2 Rat) | |
Protein A purified | |
Protein Kinase, Signal Transduction | |
3716.0 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction