Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Jak2 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$162.36 - $492.89
Specifications
| Antigen | Jak2 |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Jak2 Polyclonal specifically detects Jak2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Jak2 | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| EC 2.7.10, EC 2.7.10.2, JAK-2, Janus kinase 2Janus kinase 2 (a protein tyrosine kinase), JTK10, tyrosine-protein kinase JAK2 | |
| JAK2 | |
| IgG | |
| Affinity Purified | |
| 125 kDa |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Cell Cycle and Replication, Signal Transduction | |
| PBS with 0.02% Sodium Azide | |
| 3717 | |
| Antibody was raised against an 18 amino acid peptide near the carboxy terminus of human JAK2. The immunogen is located within the last 50 amino acids of JAK2. Amino Acid Squence: WNNNVNQRPSFRDLALR | |
| Primary | |
| JAK2 antibody is predicted to not cross-react with other members of the JAK family of proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title