Learn More
JAK2 Polyclonal Antibody, Abbomax
Rabbit Polyclonal Antibody
Supplier: Abbomax Inc 601670

Description
Positive control: Mouse Muscle Cellular location: Cytoplasmic.
JAK2 Polyclonal antibody specifically detects JAK2 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
JAK2 | |
Polyclonal | |
Unconjugated | |
JAK2 | |
EC 2.7.10.2; Fd17; JAK 2; JAK2; JAK-2; Janus kinase 2; Janus kinase 2 (a protein tyrosine kinase); JTK 10; JTK10; kinase Jak2; THCYT3; Tyrosine-protein kinase JAK2 | |
A synthetic peptide surrounding to the epitope -EYYK- without any phosphorylation of JAK2 protein | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with proprietary stabilizer and 0.01% sodium azide | |
O60674, Q62120 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16452, 3717 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.