Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JAK2 Polyclonal Antibody, PE, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS23004RPE

This item is not returnable.
View return policy
Description
JAK2 Polyclonal antibody specifically detects JAK2 in Human samples. It is validated for Flow CytometrySpecifications
JAK2 | |
Polyclonal | |
PE | |
JAK2 | |
EC 2.7.10.2; Fd17; JAK 2; JAK2; JAK-2; Janus kinase 2; Janus kinase 2 (a protein tyrosine kinase); JTK 10; JTK10; kinase Jak2; THCYT3; Tyrosine-protein kinase JAK2 | |
KLH conjugated synthetic peptide derived from human JAK2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry | |
1 mg/mL | |
TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300 | |
O60674 | |
Rabbit | |
Protein A | |
RUO | |
3717 | |
-20°C or -80°C if preferred | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction