Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Jak2 Rabbit anti-Human, Mouse, Clone: 4B0K1, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $460.50
Specifications
Antigen | Jak2 |
---|---|
Clone | 4B0K1 |
Applications | Western Blot, Immunoprecipitation |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Jak2 Monoclonal antibody specifically detects Jak2 in Human, Mouse samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
Jak2 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Cell Cycle and Replication, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
3717 | |
IgG | |
Affinity purified |
4B0K1 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
EC 2.7.10, EC 2.7.10.2, JAK-2, Janus kinase 2Janus kinase 2 (a protein tyrosine kinase), JTK10, tyrosine-protein kinase JAK2 | |
A synthetic peptide corresponding to a sequence within amino acids 1033-1132 of human Jak2 (O60674). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title