Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Jak3 Antibody (452524), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB46991J
Description
Jak3 Monoclonal antibody specifically detects Jak3 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOFSpecifications
Jak3 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF | |
452524 | |
Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
EC 2.7.10, JAK3_HUMAN, JAK-3EC 2.7.10.2, JAKLtyrosine-protein kinase JAK3, Janus kinase 3Leukocyte janus kinase, LJAK, L-JAKJanus kinase 3 (a protein tyrosine kinase, leukocyte) | |
E. coli-derived recombinant human Jak3, Gly46-Thr209, Accession # P52333 | |
0.1 mL | |
Cell Biology, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
3718 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction