Learn More
Promega JAK3 Kinase Enzyme System, 1mg
Catalog No. PRV3702 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV3702 1 mg
1 options
Catalog No. PRV3702 Supplier Promega Supplier No. V3702
Add to Cart
Add to Cart
Request a Quote
Recombinant human JAK3 (amino acids 781-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. JAK3 is a member of the JAK family of tyrosine kinases involved in cytokine receptor-mediated intracellular signal transduction.
  • Easily Screen and Profile JAK3 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C