Learn More
Invitrogen™ JAK3 Monoclonal Antibody (5H2)

Description
MA5-15561 targets JAK3 in indirect ELISA, FACS, IF, and WB applications and shows reactivity with Human and mouse samples. The MA5-15561 immunogen is purified recombinant fragment of human JAK3 expressed in E. Coli. MA5-15561 detects JAK3 which has a predicted molecular weight of approximately 125kDa.
Specifications
Specifications
| Antigen | JAK3 |
| Applications | ELISA, Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 5H2 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Ascites with 0.03% sodium azide |
| Gene | JAK3 |
| Gene Accession No. | P52333, Q62137 |
| Gene Alias | Breast-JAK3; fae; JAK3; JAK-3; JAK3_HUMAN; JAK3B; JAKL; Janus kinase 3; Janus kinase 3 (a protein tyrosine kinase, leukocyte); Janus kinase 3 protein-tyrosine kinase; Janus kinase 3, protein-tyrosine kinase; L JAK; Leukocyte janus kinase; LJAK; L-JAK; RATJAK3; Tyrosine-protein kinase JAK3; wil |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.