Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JAK3 Recombinant Rabbit Monoclonal Antibody (3N1), Proteintech
SDP

Catalog No. 502334243 Shop All Proteintech Group Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-233-4243 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-233-4243 Supplier Proteintech Group Inc Supplier No. 803311RR100UL
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

JAK3 Recombinant Monoclonal antibody specifically detects JAK3 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen JAK3
Applications Western Blot
Classification Recombinant Monoclonal
Clone 3N1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Accession No. P52333
Gene Alias JAK-3; JAK3; JAK3_HUMAN; JAKL; L-JAK; LJAK
Host Species Rabbit
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3718
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.