Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ JAKMIP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541941
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541941 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541941 Supplier Invitrogen™ Supplier No. PA541941
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: FLRLQVLEQQ HVIDDLSLER ERLLRSKRHR GKSLKPPKKH VVETFFGFDE Sequence homology: Cow: 92%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%.

JAKMIP1, also known as Marlin-1, is a highly-conserved vertebrate-specific microtubule-associated protein that plays a role in JAK1 signaling and regulate microtubule cytoskeleton rearrangements. JAKMIP1 is expressed predominantly in neuronal and lymphoid cells and colocalizes with microtubules. Multiple isoforms of JAKMIP1 are known to exist: isoforms 1 and 5 are also expressed in liver, lung and skeletal muscle, while isoforms 2-4 are specifically expressed in brain and retina. JAKMIP1 participates in polarized secretion in lymphocytes and it has been suggested to be a novel effector memory gene restraining T cell-mediated cytotoxicity. Dysregulation of JAKMIP1 gene expression is associated with genetic conditions linked to autism in humans.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JAKMIP1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene JAKMIP1
Gene Accession No. Q96N16
Gene Alias 5830437M04Rik; C330021K24Rik; FLJ31564; GABA-B receptor-binding protein; Gababrbp; gamma-aminobutyric acid (GABA-B) receptor binding protein; Jak and microtubule interacting protein 1; Jakmip1; JAMIP1; janus kinase and microtubule interacting protein 1; janus kinase and microtubule-interacting protein 1; MARLIN1; Marlin-1; Multiple alpha helices and RNA-linker protein 1; multiple alpha-helices and RNA-linker protein 1; multiple coiled-coil GABABR1-binding protein
Gene Symbols JAKMIP1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human JAKMIP1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 152789
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.