Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JAKMIP2 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2412020.025MG
Description
JAKMIP2 Polyclonal specifically detects JAKMIP2 in Human, Mouse samples. It is validated for Western Blot, ELISA.Specifications
JAKMIP2 | |
Polyclonal | |
Unconjugated | |
CTCL tumor antigen HD-CL-04, JAMIP2KIAA0555Jak and microtubule interacting protein 2, janus kinase and microtubule interacting protein 2, janus kinase and microtubule-interacting protein 2, NECC1, neuroendocrine long coiled-coil 1, Neuroendocrine long coiled-coil protein 1 | |
Rabbit | |
89 kDa | |
0.025 mg | |
Primary | |
At least four isoforms of JAKMIP2 are known to exist; this antibody will detect all four isoforms. JAKMIP2 antibody is predicted to not cross-react with JAKMIP1. | |
Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Western Blot, ELISA | |
1 mg/mL | |
PBS with 0.02% Sodium Azide | |
JAKMIP2 | |
Antibody was raised against a 16 amino acid peptide near the center of human JAKMIP2. The immunogen is located within amino acids 250 - 300 of JAKMIP2. Amino Acid Squence: RAGDGSEHCSSPDLR | |
Affinity Purified | |
RUO | |
9832 | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction