Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ JAKMIP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116582
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116582 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116582 Supplier Invitrogen™ Supplier No. PA5116582
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: A-549, 293T, HepG2, HeLa, K-562, Jurkat, Mouse brain, Rat brain Immunogen sequence: EKQEAENHRL QQELQDARDQ NELLEFRNLE LEERERRSPP FNLQIHPFSD GVSALQIYCM KEGVKDVNIP DLIKQLDILG DNGNLRNEEQ VAIIQASTVL SLAEKWIQQI EGAEAALHQK MMELESDMEQ FCKIKGYLEE ELDYRKQALD QAYMRIQELE ATLYNALQQE TVIKFGELLS EKQQEELRTA VEKLRRQMLR KSREYDCQIL QERMELLQQA HQRIRDLEDK TDIQKRQIKD LEEKSNRKHG.

JAKMIP2 (janus kinase and microtubule-interacting protein 2), also known as NECC1 (neuroendocrine long coiled-coil protein 1), CTCL tumor antigen HD-CL-04, JAMIP2 or KIAA0555, is a 810 amino acid protein belonging to the JAKMIP family. Localizing to the Golgi apparatus, JAKMIP2 is high expressed in brain, with moderate levels of expression found in thymus, spleen and lung. Existing as three alternatively spliced isoforms, the gene encoding JAKMIP2 maps to human chromosome 5q32 and mouse chromosome 18 B3. Chromosome 5 contains 181 million base pairs and comprises nearly 6% of the human genome. Deletion of the p arm of chromosome 5 leads to Cri du chat syndrome, while deletion of the q arm or of chromosome 5 altogether is common in therapy-related acute myelogenous leukemias and myelodysplastic syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JAKMIP2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.90 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Jakmip2
Gene Accession No. Q96AA8
Gene Alias 6430702L21Rik; AI850334; CTCL tumor antigen HD-CL-04; D930046L20Rik; Jak and microtubule interacting protein 2; JAKMIP2; JAMIP2; janus kinase and microtubule interacting protein 2; janus kinase and microtubule-interacting protein 2; KIAA0555; NECC1; Neuroendocrine long coiled-coil protein 1; RGD1559742
Gene Symbols Jakmip2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 561-810 of human JAKMIP2 (NP_0556052).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 307479, 76217, 9832
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.