Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ JAKMIP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561374
Description
Immunogen sequence: KLQVIEQQNI IDELTRDREK LIRRRKHRRS SKPIKRPVLD PFIGYDEDSM DSETSSMASF RTDRT Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
JAKMIP2 (janus kinase and microtubule-interacting protein 2), also known as NECC1 (neuroendocrine long coiled-coil protein 1), CTCL tumor antigen HD-CL-04, JAMIP2 or KIAA0555, is a 810 amino acid protein belonging to the JAKMIP family. Localizing to the Golgi apparatus, JAKMIP2 is high expressed in brain, with moderate levels of expression found in thymus, spleen and lung. Existing as three alternatively spliced isoforms, the gene encoding JAKMIP2 maps to human chromosome 5q32 and mouse chromosome 18 B3. Chromosome 5 contains 181 million base pairs and comprises nearly 6% of the human genome. Deletion of the p arm of chromosome 5 leads to Cri du chat syndrome, while deletion of the q arm or of chromosome 5 altogether is common in therapy-related acute myelogenous leukemias and myelodysplastic syndrome.
Specifications
JAKMIP2 | |
Polyclonal | |
Unconjugated | |
Jakmip2 | |
6430702L21Rik; AI850334; CTCL tumor antigen HD-CL-04; D930046L20Rik; Jak and microtubule interacting protein 2; JAKMIP2; JAMIP2; janus kinase and microtubule interacting protein 2; janus kinase and microtubule-interacting protein 2; KIAA0555; NECC1; Neuroendocrine long coiled-coil protein 1; RGD1559742 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9832 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96AA8 | |
Jakmip2 | |
Recombinant protein corresponding to Human JAKMIP2. Recombinant protein control fragment (Product #RP-100239). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction