Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JAM-4/IGSF5 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299581C
Description
JAM-4/IGSF5 Polyclonal specifically detects JAM-4/IGSF5 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
JAM-4/IGSF5 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
GSF5, IgSF5, Immunoglobulin Superfamily 5 Like, Immunoglobulin Superfamily Member 5, Immunoglobulin Superfamily, Member 5, JAM4, JAM-4, Junctional Adhesion Molecule 4, Putative Gene, Immunoglobulin Superfamily 5 Like11 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of mouse JAM-4/IGSF5. | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
150084 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction