Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JAM-A Antibody (H202-106), Biotin, Novus Biologicals™

Rat Monoclonal Antibody has been used in 4 publications
Supplier: Novus Biologicals NB10063716100UG
Description
JAM-A Monoclonal specifically detects JAM-A in Mouse samples. It is validated for Flow Cytometry.Specifications
JAM-A | |
Monoclonal | |
0.1 mg/mL | |
Flow Cytometry Neat-1:5 | |
CD321 antigen, F11 receptor, JAM-1, JAM1CD321, JAMA, JAM-A, JCAMJAM, Junctional adhesion molecule 1Platelet F11 receptor, junctional adhesion molecule A, PAM-1KAT, Platelet adhesion molecule 1 | |
Rat | |
100 μg | |
Mouse | |
Purified |
Flow Cytometry | |
H202-106 | |
Biotin | |
PBS (pH 7.4) and 1.0% BSA | |
F11R | |
MTE1/MTE2 stromal cell lines. | |
50848 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction