Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JAM-A Antibody (H202-106), PE, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NB10065340PE
Description
JAM-A Monoclonal specifically detects JAM-A in Mouse samples. It is validated for Flow Cytometry.Specifications
| JAM-A | |
| Monoclonal | |
| PE | |
| CD321 antigen, F11 receptor, JAM-1, JAM1CD321, JAMA, JAM-A, JCAMJAM, Junctional adhesion molecule 1Platelet F11 receptor, junctional adhesion molecule A, PAM-1KAT, Platelet adhesion molecule 1 | |
| Rat | |
| Protein G purified | |
| RUO | |
| 50848 | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| H202-106 | |
| Flow Cytometry | |
| F11R | |
| MTE1/MTE2 stromal cell lines. | |
| 0.25 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction