Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JAM-A Antibody (OTI3H3), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271876H
Description
JAM-A Monoclonal specifically detects JAM-A in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
JAM-A | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
F11R | |
Full length recombinant protein of human JAM-A (NP_653087) produced in HEK293T cells. | |
0.1 mL | |
50848.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI3H3 | |
Western Blot, Immunohistochemistry | |
CD321 antigen, F11 receptor, JAM-1, JAM1CD321, JAMA, JAM-A, JCAMJAM, Junctional adhesion molecule 1Platelet F11 receptor, junctional adhesion molecule A, PAM-1KAT, Platelet adhesion molecule 1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction