Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ JAM-A Antibody Pair [HRP]
SDP

Catalog No. NB010373 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
15 Plates
5 Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB010373 15 Plates
NB010374 5 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB010373 Supplier Novus Biologicals™ Supplier No. NBP27966715PLATES
Add to Cart
Add to Cart

Mouse Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human JAM-A.

Specifications

Assay Range 7.8 pg/mL
Assay Sensitivity 7.8-500 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human JAM-A.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias CD321 antigen, F11 receptor, JAM-1, JAM1CD321, JAMA, JAM-A, JCAMJAM, Junctional adhesion molecule 1Platelet F11 receptor, junctional adhesion molecule A, PAM-1KAT, Platelet adhesion molecule 1
Gene ID (Entrez) 50848
Gene Symbol F11R
Host Species Mouse
Kit Contents
  • Detection Antibody: 0.1 mg/mL of mouse anti-Human JAM-A monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Capture Antibody: 0.5 mg/mL of mouse anti-Human JAM-A monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 10 ng of recombinant Human JAM-A.
  • Quantity 15 Plates
    Regulatory Status RUO
    Storage Requirements Storage is content dependent.
    Target JAM-A
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.4 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 500 pg/mL is recommended.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.