Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JAM-A Antibody (RM0116-1M21), PerCP, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP122494PCP
Description
JAM-A Monoclonal antibody specifically detects JAM-A in Mouse samples. It is validated for Western BlotSpecifications
JAM-A | |
Monoclonal | |
PerCP | |
PBS | |
Rat | |
Protein A or G purified | |
RUO | |
50848 | |
Store at 4C in the dark. | |
IgG2 |
Western Blot | |
RM0116-1M21 | |
Western Blot | |
CD321 antigen, F11 receptor, JAM-1, JAM1CD321, JAMA, JAM-A, JCAMJAM, Junctional adhesion molecule 1Platelet F11 receptor, junctional adhesion molecule A, PAM-1KAT, Platelet adhesion molecule 1 | |
Mouse recombinant protein of the extracellular domain of JAM-A | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction