Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JAM-A Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501563744
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-3744 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-3744 Supplier Bethyl Laboratories, Inc Supplier No. A302891AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predict

CD321, also known as junctional adhesion molecule 1 (JAM-1), is a 32-41k D glycoprotein which shares similarites with related proteins JAM-2 and JAM-3. CD321 is a multifunctional protein that is primarily expressed by platelets, endothelial and epithelial cells. The CD321 protein co-localizes with tight junction molecules in both epithelial and endothelial cells and plays an important role in the regulation of junctional integrity and permeability. In addition, CD321is a ligand for the integrin LFA-1 and is also involved in the transmigration of leucocytes.

Specifications

Antigen JAM-A
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene F11R
Gene Accession No. Q9Y624
Gene Alias CD321, JAM, JAM1, JAMA, JCAM, KAT, PAM-1, F11R
Gene Symbols F11R
Host Species Rabbit
Immunogen Between 249 and 299
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50848
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.