Learn More
JAM-A Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302891A

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predicted to react with Mouse and Rat
CD321, also known as junctional adhesion molecule 1 (JAM-1), is a 32-41k D glycoprotein which shares similarites with related proteins JAM-2 and JAM-3. CD321 is a multifunctional protein that is primarily expressed by platelets, endothelial and epithelial cells. The CD321 protein co-localizes with tight junction molecules in both epithelial and endothelial cells and plays an important role in the regulation of junctional integrity and permeability. In addition, CD321is a ligand for the integrin LFA-1 and is also involved in the transmigration of leucocytes.Specifications
JAM-A | |
Polyclonal | |
Unconjugated | |
F11R | |
CD321, JAM, JAM1, JAMA, JCAM, KAT, PAM-1, F11R | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
50848 | |
4° C | |
Liquid |
Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.2 mg/ml | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9Y624 | |
F11R | |
Between 249 and 299 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.