Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ JAM2 Monoclonal Antibody (6G1A11)
GREENER_CHOICE

Catalog No. FERMA548774
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA548774 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA548774 Supplier Invitrogen™ Supplier No. MA548774
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

JAM2 Monoclonal Antibody for Western Blot, Flow, ELISA

Tight junctions represent one mode of cell-to-cell adhesion in epithelial or endothelial cell sheets, forming continuous seals around cells and serving as a physical barrier to prevent solutes and water from passing freely through the paracellular space. The protein encoded by this immunoglobulin superfamily gene member is localized in the tight junctions between high endothelial cells. It acts as an adhesive ligand for interacting with a variety of immune cell types and may play a role in lymphocyte homing to secondary lymphoid organs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JAM2
Applications ELISA, Flow Cytometry, Western Blot, Western Blot
Classification Monoclonal
Clone 6G1A11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene JAM2
Gene Accession No. P57087
Gene Alias 1110002N23Rik; 2410030G21Rik; 2410167M24Rik; AU016127; C21orf43; CD322; Jam2; JAM-2; JAMB; JAM-B; JAM-IT/VE-JAM; Jcam2; junction adhesion molecule 2; junction adhesion molecule B; junction cell adhesion molecule 2; junctional adhesion molecule 2; Junctional adhesion molecule B; PRO245; RP23-253E17.2; UNQ219/PRO245; Vascular endothelial junction-associated molecule; VEJAM; VE-JAM
Gene Symbols JAM2
Host Species Mouse
Immunogen Purified recombinant fragment of human CD322 (AA: extra 29-238) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58494
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.