Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ JAM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567831
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567831 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567831 Supplier Invitrogen™ Supplier No. PA567831
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Mouse and Rat samples.

Tight junctions represent one mode of cell-to-cell adhesion in epithelial or endothelial cell sheets, forming continuous seals around cells and serving as a physical barrier to prevent solutes and water from passing freely through the paracellular space. The protein encoded by this immunoglobulin superfamily gene member is localized in the tight junctions between high endothelial cells. It acts as an adhesive ligand for interacting with a variety of immune cell types and may play a role in lymphocyte homing to secondary lymphoid organs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JAM2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene JAM2
Gene Accession No. P57087, Q9JI59
Gene Alias 1110002N23Rik; 2410030G21Rik; 2410167M24Rik; AU016127; C21orf43; CD322; Jam2; JAM-2; JAMB; JAM-B; JAM-IT/VE-JAM; Jcam2; junction adhesion molecule 2; junction adhesion molecule B; junction cell adhesion molecule 2; junctional adhesion molecule 2; Junctional adhesion molecule B; PRO245; RP23-253E17.2; UNQ219/PRO245; Vascular endothelial junction-associated molecule; VEJAM; VE-JAM
Gene Symbols JAM2
Host Species Rabbit
Immunogen Synthesized peptide derived from human JAM2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58494, 619374, 67374
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.