Learn More
Invitrogen™ JAM3 (soluble) Monoclonal Antibody (18F26)
Description
MA1-40060 detects JAM-C from mouse and human samples. MA1-40060 has been successfully used in flow cytometry, ELISA, functional assays, immunohistochemistry (frozen), and immunoprecipitation applications. ELISA: Antibody CRAM-18 F26 can be used as a detection antibody. The MA1-40060 immunogen is mouse JAM-C.
Specifications
Specifications
| Antigen | JAM3 (soluble) |
| Applications | ELISA, Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Immunoprecipitation |
| Classification | Monoclonal |
| Clone | 18F26 |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% BSA and no preservative |
| Gene | JAM3 |
| Gene Accession No. | Q9BX67, Q9D8B7 |
| Gene Alias | 1110002N23Rik; JAM-2; JAM3; JAM-3; JAMC; JAM-C; Jcam3; junction adhesion molecule 3; junction cell adhesion molecule 3; junction cell adhesion molecule C; junctional adhesion molecule 3; Junctional adhesion molecule C; sJAM-C; Soluble form of JAM-C; UNQ859/PRO1868 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.