Learn More
Description
Used for: selectively staining Staphylococcus aureus.
Application: confocal microscopy and flow cytometry.
Properties and Photophysical Data: Excitation and emission maxima (λ) are 525 nm and 549 nm, respectively; quantum yield = 0.91; extinction coefficient = 122,000 M-1cm-1; lactone-zwitterion equilibrium constant (KL-Z) = 0.068.
Specifications
Specifications
| Chemical Name or Material | (R)-4-((2-Amino-2-carboxyethyl)carbamoyl)-2-(3-(3,3-difluoroazetidin-1-ium-1-ylidene)-6-(3,3-difluoroazetidin-1-yl)-3H-xanthen-9-yl)benzoate ditrifluoroacetate |
| Quantity | 2 mg |
| Product Line | Tocris |
| Target | Fluorescent Probes for Imaging Bacteria |
| Molecular Formula | C30H24F4N4O6.2CF3CO2H |
| Purity | 95% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
