Learn More
Description
Application: Suitable for flow cytometry, confocal microscopy, super resolution microscopy (SRM) including dSTORM (in both live and fixed cells) and STED. Cell permeable.
Properties and Photophysical Data: Excitation and emission maxima (λ) are 531 nm and 553 nm, respectively.
Please see the product protocol for further information and a guide to protein/antibody labeling.
Janelia Fluor is a registered trademark of Howard Hughes Medical Institute.
Specifications
Specifications
| Chemical Name or Material | 2-(3-(3,3-Difluoroazetidin-1-ium-1-ylidene)-6-(3,3-difluoroazetidin-1-yl)-3H-xanthen-9-yl)-4-((2-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)ethyl)carbamoyl)benzoate |
| Quantity | 1 mg |
| Product Line | Tocris |
| Target | Sulfhydyl-Reactive Janelia Fluor Dyes |
| Molecular Formula | C33H24F4N4O6 |
| Purity | 0.95 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
