Learn More
Description
We also offer Janelia Fluor™ conjugated antibodies and custom conjugation services with our sister company R&D Systems.
HaloTag is a trademark of Promega Corporation, and SNAP-tag is a trademark of New England BioLabs, Inc.
Specifications
Specifications
| Chemical Name or Material | 3,6-Di-1-azetidinyl-9-[2-carboxy-5-[[(2,5-dioxo-1-pyrrolidinyl)oxy]carbonyl]phenyl]xanthylium, inner salt |
| CAS | 1811539-32-8 |
| Synonym | JF 549, NHS |
| Quantity | 2 mg |
| Product Line | Tocris |
| Target | Janelia Fluor Dyes |
| Molecular Formula | C31H25N3O7 |
| Formula Weight | 551.56 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

