Learn More
Description
Application: Live-cell imaging as a self-labeling tag substrate. Suitable for confocal microscopy, light sheet microscropy, super resolution microscopy (SRM) techniques including dSTORM (in both live and fixed cells). Stable to fixation. Cell permeable.
Properties and Photophysical Data: Janelia Fluor™ 635, Haloalkane shows a high degree of fluorogenicity. Excitation and emission maxima (λ) are 635 nm and 652 nm, respectively; quantum yield = 0.56; extinction coefficient = 167,000 M-1cm-1 (measured in ethanol or TFE plus 0.1% TFA); lactone-zwitterion equilibrium constant (KL-Z) <0.0001. Turn-on ratio in vitro A/A0 = 113, F/F0 = 67. Exhibits low non-specific background staining.
Janelia Fluor is a registered trademark of Howard Hughes Medical Institute.
Specifications
Specifications
| Chemical Name or Material | 4-((2-(2-((6-Chlorohexyl)oxy)ethoxy)ethyl)carbamoyl)-2-(3-(3-fluoroazetidin-1-ium-1-ylidene)-7-(3-fluoroazetidin-1-yl)-5,5-dimethyl-3,5-dihydrodibenzo[b,e]silin-10-yl)benzoate |
| CAS | 1811539-66-8 |
| Quantity | 10 μg |
| Product Line | Tocris |
| Target | Janelia Fluor Dyes |
| Molecular Formula | C39H46ClF2N3O5Si |
| Physical Form | Solid |
| Color | White |
| Product Type | Bioactive Small Molecule |
| Purity | 0.95 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
