Learn More
Description
Spontaneously Blinking Janelia Fluor™ Dyes allow facile single-molecule localization microscopy (SMLM) in cells and dense biomolecular structures, without the need for photoactivation or redox buffers. These spontaneously blinking dyes harness Janelia Fluor™ technology to delivery dyes that automatically cycle between 'off' and 'on' states with an ideal duty cycle for super resolution microscopy experiments.
Key Information: Janelia Fluor™ 635b, Maleimide is a spontaneously blinking fluorescent dye, supplied with a maleimide reactive group for conjugation (thiol reactivity) for the labeling of protein.
Used for: suitable to characterize the physical properties of dense biomolecule condensates and complex macromolecular assemblies.
Application: suitable for super resolution microscopy (SRM) including single-molecule localization microscopy (SMLM).
Properties and Photophysical Data: derived from Janelia Fluor™ 635, Maleimide (Cat. No. 8027). Excitation and emission maxima (λ) are 651 nM and 667 nM, respectively.
Janelia Fluor is a registered trademark of Howard Hughes Medical Institute.
Specifications
Specifications
| Chemical Name or Material | N-(2-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)ethyl)-3,7-bis(3-fluoroazetidin-1-yl)-5,5-dimethyl-3'H,5H-spiro[dibenzo[b,e]siline-10,1'-isobenzofuran]-6'-carboxamide |
| Quantity | 200 μg |
| Product Line | Tocris |
| Target | Sulfhydyl-Reactive Janelia Fluor Dyes |
| Molecular Formula | C35H34F2N4O4Si |
| Purity | 0.9 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

