Learn More
Description
Key Information: Janelia Fluor™ 635b, NHS ester is a spontaneously blinking fluorescent dye, supplied with an NHS ester reactive group for the labeling of primary amines.
Used for: suitable for DNA labelling using amine-terminated oligonucleotides. Also used to label oligonucleotides for super-resolution assays of transposase-accessible chromatin (ATAC-SMLM).
Application: suitable for super resolution microscopy (SRM) including single-molecule localization microscopy (SMLM).
Properties and Photophysical Data: derived from Janelia Fluor™ 635, SE (Cat. No. 6419). Excitation and emission maxima (λ) are 654 nM and 671 nM, respectively.
Janelia Fluor is a registered trademark of Howard Hughes Medical Institute.
Specifications
Specifications
| Chemical Name or Material | 2,5-Dioxopyrrolidin-1-yl 3,7-bis(3-fluoroazetidin-1-yl)-5,5-dimethyl-3'H,5H-spiro[dibenzo[b,e]siline-10,1'-isobenzofuran]-6'-carboxylate |
| Quantity | 200 μg |
| Product Line | Tocris |
| Target | Amine-Reactive Janelia Fluor Dyes |
| Molecular Formula | C33H31F2N3O5Si |
| Purity | 0.9 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

