Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Janelia Fluor 646, free acid

Catalog No. 69935
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-935 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 69-935 Supplier Tocris Bioscience™ Supplier No. 6993/5
This item is not returnable. View return policy

Red dye (Reactive group: free acid). Applications: super resolution microscopy e.g. dSTORM & STED, confocal, flow and live cell imaging

Janelia Fluor® 646, free acid is a red fluorogenic fluorescent dye; supplied as a free acid. Used in protocol (2017 Grimm et al - see references below) for the synthesis of Janelia Fluor™ HaloTag™ and SNAP-Tag™ ligands, for use in live cell imaging experiments. Suitable for confocal fluorescent imaging, super resolution microscopy (SRM) techniques such as dSTORM (live and fixed cells) and STED imaging. Also suitable for flow cytometry. Cell permeable. Excitation maximum = 646 nm; emission maximum = 664 nm; Quantum yield = 0.54; Extinction coefficient = 152,000 M-1cm-1.

To measure the absorbance spectrum of this dye we recommend the following solvent: ethanol or trifluoroethanol plus 0.1% TFA.

We also offer Janelia Fluor™ conjugated antibodies and custom conjugation services with our sister company Novus Biologicals.

HaloTag is a trademark of Promega Corporation, and SNAP-tag is a trademark of New England BioLabs, Inc.

Specifications

Chemical Name or Material 1-[7-(1-Azetidinyl)-10-(2,5-dicarboxyphenyl)-9,9-dimethyl-9-silaanthracen-2(9 H)-ylidene]azetidinium inner salt trifluoroacetate
Quantity 5 mg
Target Free Acid Janelia Fluor™ Dyes
Molecular Formula C29H28N2O4Si.CF3CO2H
Formula Weight 3737
Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.