Learn More
Tocris Bioscience™ Janelia Fluor 646, Maleimide
Description
To measure the absorbance spectrum of this dye we recommend the following solvent: ethanol or trifluoroethanol plus 0.1% TFA.
We also offer Janelia Fluor™ conjugated antibodies and custom conjugation services with our sister company Novus Biologicals.
Specifications
Specifications
| Chemical Name or Material | 2-(3-(Azetidin-1-ium-1-ylidene)-7-(azetidin-1-yl)-5,5-dimethyl-3,5-dihydrodibenzo[b,e]silin-10-yl)-4-((2-(2,5-dioxo-2,5-dihydro-1 H-pyrrol-1-yl)ethyl)carbamoyl)benzoate |
| Quantity | 2 mg |
| Target | Sulfhydyl-Reactive Janelia Fluor™ Dyes |
| Molecular Formula | C35H34N4O5Si |
| Formula Weight | 618.75 |
| Purity | 95% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.