Learn More
Description
Used for: imaging Mycobacterium tuberculosis in TB drug susceptibility testing.
Application: fluorescent microscopy and flow cytometry.
Properties and Photophysical Data: Excitation and emission maxima (λ) are 646 nm and 664 nm, respectively; quantum yield = 0.54; extinction coefficient = 152,000 M-1cm-1 (measured in ethanol plus 0.1% TFA); A280 correction factor = 0.19; lactone-zwitterion equilibrium constant (KL-Z) = 0.0012.
Janelia Fluor™ is a registered trademark of Howard Hughes Medical Institute.
Specifications
Specifications
| Chemical Name or Material | 2-(3-(Azetidin-1-ium-1-ylidene)-7-(azetidin-1-yl)-5,5-dimethyl-3,5-dihydrodibenzo[b,e]silin-10-yl)-4-((((2R,3S,4S,5R,6R)-3,4,5-trihydroxy-6-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)methyl)c |
| Quantity | 2 mg |
| Product Line | Tocris |
| Target | Fluorescent Probes for Imaging Bacteria |
| Molecular Formula | C41H49N3O13Si |
| Purity | 95% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
