Learn More
Description
Application: Suitable for flow cytometry, confocal microscopy, super resolution microscopy (SRM) including dSTORM (in both live and fixed cells) and STED. Can be multiplexed for two color imaging with Janelia Fluor™ 549 SE (Cat. No. 6147). Cell permeable.
Properties and Photophysical Data: Excitation and emission maxima (λ) are 669 nm and 689 nm, respectively.
Please see the product protocol for further information and a guide to protein/antibody labeling.
To measure the absorbance spectrum of this dye we recommend the following solvent: ethanol or trifluoroethanol plus 0.1% TFA.
We also offer Janelia Fluor™ conjugated antibodies and custom conjugation services with our sister company R&D Systems.
Janelia Fluor is a registered trademark of Howard Hughes Medical Institute.
Specifications
Specifications
| Chemical Name or Material | 2-(3-(Azetidin-1-ium-1-ylidene)-7-(azetidin-1-yl)-5,5-dimethyl-3,5-dihydrodibenzo[b,e]silin-10-yl)-4-((2-((2-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)ethyl)amino)-2-oxoethyl)thio)-3,5,6-trifluorobenzoate |
| Quantity | 200 μg |
| Product Line | Tocris |
| Target | Fluorescent Dyes |
| Molecular Formula | C36H33F3N4O5SSi |
| Purity | 0.95 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
